Test kits, photometric, Carbonate/carbon dioxide, LCK cuvette tests
705-3039
: Hach
:
- Chemical method:pH Indicator
- Pk:25 Tests
- Description:Carbonate/Carbon dioxide
- Seq:210
- Type:LCK388
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:55 - 550 mg/l CO₂
Ready for use, photometric evaluation system and environmentally friendly.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C