Test kits, photometric, Cadmium, LCK cuvette tests
Catalog # 705-3016
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Cadion
- Pk:25 Tests
- Description:Cadmium
- Seq:209
- Type:LCK308
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:0,02 - 0,3 mg/l Cd
Specifications
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)