Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium carbonate
Calcium carbonate Ph. Eur., Macron Fine Chemicals™
Calcium carbonate Ph. Eur., Macron Fine Chemicals™
Calcium carbonate Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
Calcium carbonate Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:
471-34-1
:
Formula:
CaCO₃
MDL Number:
MFCD00010906
:
100.09 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
SDS
sds_MACRP104
Documentation