Organic reference standards, dacarbazine, Ph. Eur.
Dacarbazine impurity B, 5-amino-1H-imidazole-4-carboxamide, Ph. Eur. standard, Reference standard, 10 mg
Catalog # EDQMY0000735
Supplier: EDQM
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:10 mg
- Standards:Reference standard
- Description:Dacarbazine impurity B, 5-amino-1H-imidazole-4-carboxamide, Ph. Eur. standard
- Cat. No.:EDQMY0000735
- Formula:C₄H₆N₄O
- Description:Dacarbazine impurity B, 5-amino-1H-imidazole-4-carboxamide
- CAS No.:360-97-4



