Order Entry
Export
ContactUsLinkComponent
Anti-Thy1.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Anti-Thy1.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Catalog # ABNOMAB15492
Supplier:  Abnova
Anti-Thy1.2 Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 30-H12]
Catalog # ABNOMAB15492
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Thy1.2
  • Clonality:
    Monoclonal
  • Clone:
    30-H12
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light.
  • Immunogen:
    Mouse thymocytes and splenocytes.
  • Purification:
    Affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal antibody raised against native mouse Thy1.2.

Type: Primary
Antigen: Thy1.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse