Anti-Thy1.2 Rat Monoclonal Antibody (PerCP) [clone: 30-H12]
Catalog # ABNOMAB15490
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Thy1.2
- Clonality:Monoclonal
- Clone:30-H12
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Form:Liquid
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:Mouse thymocytes and splenocytes.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against native mouse Thy1.2.
Type: Primary
Antigen: Thy1.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PerCP
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse