Anti-S100A1 Mouse Monoclonal Antibody [clone: S1/61]
ABNOMAB14975
: Abnova
- Antibody type:Primary
- Antigen symbol:S100A1
- Clonality:Monoclonal
- Clone:S1/61
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:6271
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human S100 (conjugated with methylated BSA).
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against native human S100A1.
Type: Primary
Antigen: S100A1
Clonality: Monoclonal
Clone: S1/61
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human