Order Entry
Export
ContactUsLinkComponent
Anti-Ptprc Rat Monoclonal Antibody (PerCP) [clone: 30-F11]
Anti-Ptprc Rat Monoclonal Antibody (PerCP) [clone: 30-F11]
Catalog # ABNOMAB15482
Supplier:  Abnova
Anti-Ptprc Rat Monoclonal Antibody (PerCP) [clone: 30-F11]
Catalog # ABNOMAB15482
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase, receptor type C
  • Antigen symbol:
    Ptprc
  • Clonality:
    Monoclonal
  • Clone:
    30-F11
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    19264
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light.
  • Immunogen:
    Mouse thymocytes and splenocytes.
  • Purification:
    Affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal antibody raised against native mouse Ptprc.

Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PerCP
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse