Anti-IPO-38 Mouse Monoclonal Antibody [clone: SPM515]
Catalog # ABNOMAB15167
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:IPO-38
- Clonality:Monoclonal
- Clone:SPM515
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at 4 °C. For long term storage store at -20 °C to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Spleen cells of a patient with hairy cell leukemia.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against human IPO-38.
Type: Primary
Antigen: IPO-38
Clonality: Monoclonal
Clone: SPM515
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human