Anti-H2-D1 Mouse Monoclonal Antibody (PerCP) [clone: 34-2-12]
ABNOMAB15449
: Abnova
- Antibody type:Primary
- Antigen name:H2-D1 molecule
- Antigen symbol:H2-D1
- Clonality:Monoclonal
- Clone:34-2-12
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:14964
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:(C57BL/6 x DBA/2) F1 mouse splenocytes.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against native mouse H2-D1.
Type: Primary
Antigen: H2-D1
Clonality: Monoclonal
Clone: 34-2-12
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse