Anti-CGA Mouse Monoclonal Antibody [clone: HCGab/52]
Catalog # ABNOMAB14713
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:hCG protein
- Antigen symbol:CGA
- Clonality:Monoclonal
- Clone:HCGab/52
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:1081|1082
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at 4 °C. For long term storage store at -20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified hCG protein.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against intact-HCG.
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: HCGab/52
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human