Eyewash station and box
Supplier: Plum Safety
Certificates
About this item
Available as either an open station or closed, dust-proof wall box containing 200 ml pH neutral and 500 ml Plum eyewash. Ideal where there is a risk of eye injuries from acids or alkalis.
- Open station with mirror and pictogram
- Wall box with mirror and eyewash sign
- Wall box ideal for dusty and dirty production areas
Rinsing time: ~2 minutes for pH neutral, 5 minutes for eyewash, shelf life: 3 years.
W×D×H open station: 230×80×290 mm, W×D×H wall box: 227×110×265 mm.