Anti-CD5 Rat Monoclonal Antibody (Biotin) [clone: 53-7.3]
Catalog # ABNOMAB15311
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:53-7.3
- Conjugation:Biotin
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12507
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4°C.
- Immunogen:Mouse thymocytes/splenocytes.
- Purification:Affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against mouse Cd5.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse