Order Entry
Export
ContactUsLinkComponent
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.72]
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.72]
Catalog # ABNOMAB15299
Supplier:  Abnova
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.72]
Catalog # ABNOMAB15299
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8a molecule
  • Antigen symbol:
    CD8a
  • Clonality:
    Monoclonal
  • Clone:
    53-6.72
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    12525
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4°C.
  • Immunogen:
    Mouse thymocytes/splenocytes.
  • Purification:
    Affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal antibody raised against mouse Cd8a.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.72
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse