Anti-TFRC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FG2/12]
Catalog # ABNOMAB15286
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transferrin receptor protein 1
- Antigen symbol:TFRC
- Clonality:Monoclonal
- Clone:FG2/12
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:7037
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human TFRC.
- Purification:Affinity chromatography
- Size:100 Reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human TFRC.
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: FG2/12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human