Orders LinkContactUsLinkComponent
Calorimetric standards
Supplier:   PARR
Calorimetric standards
Supplier:  PARR
Certificates
Calorimetric standards
3 Options Available Below
Product Details & Documents
Calorific standard in powder or pellet form for standardising all oxygen bomb calorimeters.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • PARR3415
    Benzoic acid, 1,0 gram pellets, bottle of 100
    Benzoic acid, 1,0 gram pellets, bottle of 100
    1
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
  • PARR3416
    Benzoic acid, 1,0 gram pellets, bottle of 500
    Benzoic acid, 1,0 gram pellets, bottle of 500
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • PARR3417
    Benzoic acid, 2,0 gram pellets, bottle of 500
    Benzoic acid, 2,0 gram pellets, bottle of 500
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g