Corning® Trypsin
6
Supplier: Corning
Jump to:
Product Details & Documents
0,25% Trypsin in HBSS (Hank’s Balanced Salt Solution) without calcium and magnesium.
- Porcine parvovirus tested
- 6×100 ml
Trypsin is an enzymatic reagent that is used to release adherent cells from tissue plates for passaging. This offering contains trypsin at a concentration of 0,25%.
Documents
Specifications
Specifications
- Pk:6
- Cat. No.:25-050-CI
- Enzyme name:Trypsin
- CAS No.:9002-07-7



