Specifications
- Antibody type:Primary
- Antigen name:Immunoglobin superfamily, member 21
- Antigen symbol:IGSF21
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:84966
- Antigen synonyms:FLJ41177|IGSF 21|IGS21_HUMAN.|immunoglobulin superfamily member 21|Immunoglobin superfamily member 21|RP11 121A23.1|MGC15730
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
IGSF21 is a 467 amino acid secreted protein that contains two Ig-like (immunoglobulin-like) domains and is a member of the immunoglobulin superfamily. Members of this family of proteins usually localize to the cell membrane, and may act as receptors in the immune response pathways. The gene encoding IGSF21 maps to human chromosome 1p36.13 and mouse chromosome 4 D3. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. A large number of disease-associated genes are associated with chromosome 1, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IGSF21
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat