Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 16
- Antigen symbol:RNF16
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:51127
- Antigen synonyms:Testis RING finger protein|RBCC|Tripartite mot protein 17|RING finger protein 16|TERF|TRIM 17|RNF16|TRI17_MOUSE.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The tripartite motif (TRIM) family of proteins are characterized by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. TRIM17 (tripartite motif-containing 17), also known as RBCC, terf or RNF16, is a 477 amino acid protein that contains one RING-type zinc finger, one SPRY domain and one B box-type zinc finger. Expressed nearly exclusively in testis, TRIM17 belongs to the TRIM family and, based on its functional domains, may play a role in transcriptional regulation events. The gene encoding TRIM17 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF16
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human