Order Entry
Export
ContactUsLinkComponent
Anti-C10ORF27 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-C10ORF27 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-9720R-CY5.5
Supplier:  Bioss
Anti-C10ORF27 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-9720R-CY5.5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromosome 10 open reading frame 27
  • Antigen symbol:
    C10ORF27
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

C10orf27, also known as spatial, is a 351 amino acid cytoplasmic protein belonging to the spatial family. C10orf27 is suggested to play a role in spermatid differentiation. Existing as two alternatively spliced isoforms, C10orf27 is widely expressed in multiple tissues, including brain, thymus and testis. C10orf27 may be associated with multiple sclerosis (MS) susceptibility and pathogenesis. MS is an inflammatory disease that causes gradual destruction of myelin in the central nervous system. C10orf27 is encoded by a gene located on human chromosome 10, which contains over 800 genes and 135 million nucleotides, making up nearly 4.5% of the human genome. PTEN is an important tumor suppressor gene located on chromosome 10 and, when defective, causes a genetic predisposition to cancer development known as Cowden syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C10ORF27
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat