Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S C2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5682
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Ubiquitin-dependent proteolysis mediates selective destruction of various cell cycle regulators, transcription factors and tumor suppressors. In eukaryotic cells, selective breakdown of cellular proteins is ensured by their ubiquitination and subsequent degradation by the 26S proteasome. At specific stages of development, embryo- and tissue-specific components of the 26S proteasome form, facilitating proteolysis. 20S Proteasome ?, also designated macropain subunit C2 or PROS-30, is a prosomal protein involved in a non-lysosomal ATP/ubiquitin-dependent proteolytic pathway. The entire proteasome is composed of at least 15 non-identical subunits which form a highly-ordered ring-shaped structure.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Proteasome 20S C2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat