Specifications
- Antibody type:Primary
- Antigen name:Chromosome X Open Reading Frame 56
- Antigen symbol:CXORF56
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:63932
- Antigen synonyms:CXorf56|UPF0428 protein CXorf56
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:120-170/222
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The X and Y chromosomes are the human sex chromosomes. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes. The combination of an X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number of conditions related to an unsual number and combination of sex chromosomes being inherited. More than one copy of the X chromosome with a Y chromosome causes Klinefelter's syndrome. A single copy of X alone leads to Turner's syndrome. More than 2 copies of the X chromosome, in the absence of a Y chromosome, is known as Triple X syndrome. Color blindness, hemophilia, and Duchenne muscular dystrophy are well known X chromosome-linked conditions which affect males more frequently as males carry a single X chromosome. The CXorf56 gene product has been provisionally designated CXorf56 pending further characterization.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CXORF56
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: