Order Entry
Export
ContactUsLinkComponent
Anti-TSPAN5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-TSPAN5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-9414R-HRP
Supplier:  Bioss
Anti-TSPAN5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-9414R-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tetraspanin 5
  • Antigen symbol:
    TSPAN5
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10098
  • Antigen synonyms:
    tetraspanin 5|NET-4|tetraspan 5|transmembrane 4 superfamily member 8|TSPAN-5|transmembrane 4 superfamily member 9|tetraspanin-5|TM4SF9|tetraspan NET-4|tetraspan TM4SF
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

NET-4, also known as TSPAN5 or TM4SF9, is a 268 amino acid multi-pass membrane protein that belongs to the tetraspanin family and is thought to play a role in signal transduction events related to cell development, activation, growth and motility. The gene encoding NET-4 maps to human chromosome 4, which encodes nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TSPAN5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat