Anti-TSPAN3 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-9412R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tetraspanin 3
- Antigen symbol:TSPAN3
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10099
- Antigen synonyms:TM4-A|TSPAN-3|TM4SF8
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TSPAN3 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPAN3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat