Specifications
- Antibody type:Primary
- Antigen name:F-Box and Leucine-rich Repeat Protein 21
- Antigen symbol:FBXL21
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26223
- Antigen synonyms:FBXL3B|FBXL3P|Fbl21|FBL3B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FBL3B is a 434 amino acid protein encoded by the human gene FBXL21. FBL3B contains one 40 amino acid F-box region, making it a member of the F-box family. FBL3B also contains three LRR (leucine-rich) repeats. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. F-box proteins are members of a large family that regulates cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, IkB-a and b-catenin, for degradation by the proteasome after ubiquitination. FBL3B is a substrate-recognition component of the SCF complex that interacts with Skp1 p19 and CUL-1. FBL3B is also associated with expression and regulation of circadian and cryptochrome proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FBXL21
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat