Order Entry
Export
ContactUsLinkComponent
Anti-FAM21C Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-FAM21C Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8214R-A488
Supplier:  Bioss
Anti-FAM21C Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8214R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Family With Sequence Similarity 21, Member C
  • Antigen symbol:
    FAM21C
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    253725
  • Antigen synonyms:
    FA21C_HUMAN.|FAM21A|Hypothetical protein LOC253725|Family with sequence similarity 21 member C|KIAA0592|Protein FAM21C
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Playing a essential role in the fission of tubules that serve as transport intermediates during endosome sorting, the WASH complex is present at the surface of endosomes and functions to recruit and activate the Arp2/3 complex for induction of actin polymerization. FAM21, Putative WASH complex subunit FAM21, is a component of the WASH complex. Having undergone evolutionary duplication, four highly homologous family members exist including FAM21A, FAM21B, FAM21C and FAM21D. FAM21 links the WASH complex to endosomes and is required for WASM-dependent retromer-mediated sorting. Also, by directly interacting with CapZ, FAM21 inhibits its anti-capping activity, thereby regulating actin dynamics.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM21C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat