Order Entry
Export
ContactUsLinkComponent
Anti-DSCC1 Rabbit Polyclonal Antibody (Cy5®)
Anti-DSCC1 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-7720R-CY5
 :  Bioss
Anti-DSCC1 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-7720R-CY5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    DNA replication and sister chromatid cohesion 1
  • Antigen symbol:
    DSCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    79075
  • Antigen synonyms:
    DCC1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Loads PCNA onto primed templates regulating velocity, spacing and restart activity of replication forks. May couple DNA replication to sister chromatid cohesion through regulation of the acetylation of the cohesin subunit SMC3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSCC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat