Anti-TP53I11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-7091R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tumor Protein 53-inducible Protein 11
- Antigen symbol:TP53I11
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9537
- Antigen synonyms:P5I11_HUMAN.|PIG11|p53 induced protein|Tumor protein p53 inducible protein 11|p53 induced gene 11 protein|PIG 11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TP53I11 (Tumor protein p53 inducible protein 11), also referred to as PIG11 (p53-induced gene 11), is an early transcriptional target of tumor suppressor p53. It is up-regulated in the induction of apoptosis or cell growth inhibition by multiple chemopreventive agents. However, its biological role remains unclear.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TP53I11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat