Anti-PDCD7 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-6834R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Programmed cell death protein 7
- Antigen symbol:PDCD7
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10081
- Antigen synonyms:PDCD7_HUMAN|Programmed cell death protein 7.|Pdcd7|HES18|Programmed cell death 7|MGC22015|Apoptosis-related protein ES18|ES18
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a 59 kDa protein that is associated with the U11 small nuclear ribonucleoprotein (snRNP), which is a component of the minor U12-type spliceosome responsible for catalyzing pre-mRNA splicing of U12-type introns. [provided by RefSeq, Dec 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDCD7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat