Order Entry
Export
ContactUsLinkComponent
Anti-LIN28 Rabbit Polyclonal Antibody (Cy7®)
Anti-LIN28 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6446R-CY7
Supplier:  Bioss
Anti-LIN28 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6446R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lin-28 homolog A
  • Antigen symbol:
    LIN28
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    389421, 79727
  • Antigen synonyms:
    RNA-binding proteináLIN-28|Zinc finger CCHC domain-containing protein 1|Lin-28 homolog A (C. elegans)|CSDD1|ZCCHC1|LIN28A
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    70-120/250
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Acts as a suppressor of microRNA (miRNA) biogenesis by specifically binding the precursor let-7 (pre-let-7), a miRNA precursor. Acts by binding pre-let-7 and recruiting ZCCHC11/TUT4 uridylyltransferase, leading to the terminal uridylation of pre-let-7. Uridylated pre-let-7 miRNAs fail to be processed by Dicer and undergo degradation. Specifically recognizes the 5'-GGAG-3' motif in the terminal loop of pre-let-7. Also recognizes and binds non pre-let-7 pre-miRNAs that contain the 5'-GGAG-3' motif in the terminal loop, leading to their terminal uridylation and subsequent degradation. Mediates MYC-mediated let-7 repression. Isoform 1, when overexpressed, stimulates growth of the breast adenocarcinoma cell line MCF-7. Isoform 2 has no effect on cell growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIN28
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat