Anti-ABCC10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5761R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, Subfamily C (CFTR/MRP), Member 10
- Antigen symbol:ABCC10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:89845
- Antigen synonyms:SIMRP7|EST182763|MRP7|ATP binding cassette subfamily C member 10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi ATP-dependent transporter probably involved in cellular detoxification through lipophilic anion extrusion.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABCC10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat