Anti-DPH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5714R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Diphthamide biosynthesis 1
- Antigen symbol:DPH1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1801
- Antigen synonyms:DPH2L1|OVCA1|DPH2L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
OVCA1 is required for the first step in the synthesis of diphthamide, a unique posttranslationally modified histidine found only in translation elongation factor-2 (EEF2; MIM 130610). This modification is conserved from archaebacteria to humans and serves as the target for ADP-ribosylation and inactivation of EEF2 by diphtheria toxin (DT) and Pseudomonas exotoxin A. OVCA1 is one of several enzymes involved in synthesis of diphthamide in EEF2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DPH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat