Anti-HTR3E Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-5328R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:5-Hydroxytryptamine (Serotonin) Receptor 3E, Ionotropic
- Antigen symbol:HTR3E
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:285242
- Antigen synonyms:5-HT3c1|5-HT3E|5-HT3-E
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
HTR3C belongs to the ligand-gated ion channel receptor superfamily. HTR3E is a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HTR3E
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human