Order Entry
Export
ContactUsLinkComponent
Anti-DDX58 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-DDX58 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5300R-A488
Supplier:  Bioss
Anti-DDX58 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5300R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DEAD (Asp-Glu-Ala-Asp) box polypeptide 58
  • Antigen symbol:
    DDX58
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23586
  • Antigen synonyms:
    Probable ATP-dependent RNA helicase DDX58|DEAD box protein 58|RIGI|RIG-I|RIG-1|RLR-1
  • Modification:
    phospho-Thr170
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The innate immune system detects viral infection by recognizing various viral components and triggers antiviral responses. Like the toll-like receptor 3 (TLR3), the cytoplasmic helicase retinoic acid inducible gene protein 1 (RIG1/DDX58) recognizes double-stranded (ds) RNA, a molecular pattern associated with viral infection. Unlike TLR3 however, RIG1/DDX58 activates the kinases TBK1 and IKKe through the adaptor protein IPS1. These kinases then phosphorylate the transcription factors IRF3 and IRF7 which are essential for the expression of type-I interferons. RIG1/DDX58 is required for the production of interferons in response to RNA viruses including paramyxoviruses, influenza virus, and Japanese encephalitis virus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse