Anti-CYP46A1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5046R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 46, Subfamily A, Polypeptide 1
- Antigen symbol:CYP46A1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10858
- Antigen synonyms:CYP46A1|Cholesterol 24-hydroxylase|CH24H|CYP46
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The Cyp46 gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is expressed exclusively in the brain, where it converts cholesterol to 24S-hydroxycholesterol by adding a hydroxyl group to cholesterol, producing a product that is more soluble than cholesterol and able to be exported from the brain.Cyp46 is also known as 24S-cholesterol hydroxylase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CYP46A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: