Anti-ICAM1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4616R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Intercellular Adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:3383
- Antigen synonyms:ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:240-290/532
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). During leukocyte trans-endothelial migration, ICAM1 engagement promotes the assembly of endothelial apical cups through ARHGEF26/SGEF and RHOG activation. In case of rhinovirus infection acts as a cellular receptor for the virus.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human