Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-EIF4E Rabbit Polyclonal Antibody (Cy5.5®)

Anti-EIF4E Rabbit Polyclonal Antibody (Cy5.5®)

Catalog # BOSSBS-4979R-CY5.5
Supplier:   Bioss
Anti-EIF4E Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4979R-CY5.5
Supplier:  Bioss
Anti-EIF4E Rabbit Polyclonal Antibody (Cy5.5®)
Price
960,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Clonality:
    Polyclonal
  • Purification:
    Purified by Protein A
  • Antibody type:
    Primary
  • Form:
    liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Store at 4°C for 12 months
  • Host:
    Rabbit
  • Concentration:
    1 μg/μl
  • Cross adsorption:
    No
  • Shipping temperature:
    4°C
  • Antigen name:
    eukaryotic translation initiation factor 4E
  • Cat. No.:
    BOSSBS-4979R-CY5.5
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    1977
  • Antigen synonyms:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
  • Immunogen:
    170-215/217
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Cy5.5®
  • Antigen symbol:
    EIF4E
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.