Anti-CD74 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4298R-CY5
Supplier: Bioss
Jump to:
Product Details & Documents
Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to the endosomal/lysosomal system where the antigen processing and binding of antigenic peptides to MHC class II takes place. Serves as cell surface receptor for the cytokine MIF.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Documents
bs-4298R-CY5
Datasheet
Specifications
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Store at 4°C for 12 months
- Host:Rabbit
- Concentration:1 μg/μl
- Cross adsorption:No
- Shipping temperature:4°C
- Antigen name:CD74 molecule, major histocompatibility complex, class II invariant chain
- Cat. No.:BOSSBS-4298R-CY5
- Reactivity:Human|Rat|Mouse
- Gene ID:972
- Antigen synonyms:HLADG|II|DHLAG|Ia-GAMMA
- Immunogen:249-290/296
- ImmunoChemistry:Yes
- Conjugation:Cy5®
- Antigen symbol:CD74
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.



