Anti-HTR3B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-4289R-A350
: Bioss
- Antibody type:Primary
- Antigen name:5-Hydroxytryptamine (serotonin) Receptor 3B
- Antigen symbol:HTR3B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9177
- Antigen synonyms:5-HT3B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:130-180/441
- Purification:Purified by Protein A
- Pk:100 µl
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HTR3B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat