Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein beta 8
- Antigen symbol:HSPB8
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26353
- Antigen synonyms:DHMN2|E2IG1|heat shock 27kDa protein 8|PP1629|small stress protein-like protein HSP22|heat shock protein beta-8|HMN2A|HSP22|alpha-Crystallin C chain|HMN2|protein kinase H11|E2-induced gene 1 protein|H11|CRYAC|CMT2L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Hsp22 (HSPB8) is a 196-amino acid protein that contains a central portion homologous to a highly conserved HSP-alpha crystallin domain common to all the small heat shock protein (HSP20) family members. Hsp22 is a monomeric protein which interacts with HSPB1. It displays temperature-dependent chaperone activity. The highest abundance of Hsp22 is in skeletal muscle, heart, and placenta. Mutations in the HSPB8 gene are associated with the inherited peripheral neuropathies, autosomal dominant distal hereditary motor neuropathy type IIA (dSMA) and axonal Charcot-Marie-Tooth disease type 2L (CMT2L).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSPB8
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat