Specifications
- Antibody type:Primary
- Antigen name:Surfactant protein A
- Antigen symbol:SP-A
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Surfactant Protein A|Pulmonary surfactant associated protein|SFTPA1B|pulmonary surfactant-associated glycoprotein A|SP-A2|SFPA2_MOUSE|Pulmonary surfactant-associated protein A2 precursor|SFTPA1|SP A|SP-A|SFTP1|PSP-A|Alveolar proteinosis protein|SFTPA|COLEC4|Pulmonary surfactant apoprotein|Surfactant pulmonary associated protein A1|Collectin-5.|PSPA|35 kDa pulmonary surfactant-associated protein|MGC133365|SP A1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication. [provided by RefSeq, Sep 2009].
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: SP-A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat