Anti-FUS Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-2980R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Fused in Sarcoma
- Antigen symbol:FUS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2521
- Antigen synonyms:TLS|Translocated in liposarcoma protein|FUS|Pigpen|POMp75|Fusion (involved in t(12,16) in malignant liposarcoma)
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:463-513/526
- Purification:Purified by Protein A
- Pk:100 µl
Binds both single-stranded and double-stranded DNA and promotes ATP-independent annealing of complementary single-stranded DNAs and D-loop formation in superhelical double-stranded DNA. May play a role in maintenance of genomic integrity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat