Anti-SLC37A4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-4039R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Solute Carrier Family 37 Member A4
- Antigen symbol:SLC37A4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2542
- Antigen synonyms:G6PT3|TRG-19|GSD1d|G6PT2|G6PT1|TRG19|GSD1b|PRO0685|GSD1c
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms a complex with glucose-6-phosphatase which is responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC37A4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat