Order Entry
Export
ContactUsLinkComponent
Anti-SLC37A4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-SLC37A4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-4039R-A488
Supplier:  Bioss
Anti-SLC37A4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-4039R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 37 Member A4
  • Antigen symbol:
    SLC37A4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2542
  • Antigen synonyms:
    G6PT3|TRG-19|GSD1d|G6PT2|G6PT1|TRG19|GSD1b|PRO0685|GSD1c
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms a complex with glucose-6-phosphatase which is responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC37A4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat