Order Entry
Export
ContactUsLinkComponent
Anti-PDHA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-PDHA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-4034R-A350
 :  Bioss
Anti-PDHA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-4034R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate dehydrogenase (lipoamide) alpha 1
  • Antigen symbol:
    PDHA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5160
  • Antigen synonyms:
    PDHE1-A type I|Pyruvate dehydrogenase E1 component subunit alpha|PDHCE1A|PDHAD|mitochondrial|PHE1A|PDHA|somatic form|PDHA1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase(E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex. Mutations in this gene are associated with pyruvate dehydrogenase E1-alpha deficiency and X-linked Leigh syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDHA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat