Anti-EIF4EBP1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3018R-CY5
Supplier: Bioss
Jump to:
Product Details & Documents
Regulates eIF4E activity by preventing its assembly into the eIF4F complex: hypophosphorylated form competes with EIF4G1/EIF4G3 and strongly binds to EIF4E, leading to repress translation. Mediates the regulation of protein translation by hormones, growth factors and other stimuli that signal through the MAP kinase and mTORC1 pathways.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EIF4EBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-3018R-CY5
Datasheet
Specifications
- Clonality:Polyclonal
- Modification:phospho-Ser65
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Store at 4°C for 12 months
- Host:Rabbit
- Concentration:1 μg/μl
- Cross adsorption:No
- Shipping temperature:4°C
- Antigen name:eukaryotic translation initiation factor 4E binding protein 1
- Cat. No.:BOSSBS-3018R-CY5
- Reactivity:Human|Rat|Mouse
- Gene ID:13685
- Antigen synonyms:PHAS-I|BP-1|4EBP1|4E-BP1
- Immunogen:56-80/118
- ImmunoChemistry:Yes
- Conjugation:Cy5®
- Antigen symbol:EIF4EBP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.



