Order Entry
Export
ContactUsLinkComponent
Anti-CAB39 Rabbit Polyclonal Antibody (Cy5®)
Anti-CAB39 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3636R-CY5
Supplier:  Bioss
Anti-CAB39 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3636R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium Binding Protein 39
  • Antigen symbol:
    CAB39
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51719
  • Antigen synonyms:
    MO25|CGI-66
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Mouse protein 25 alpha (MO25 alpha, CAB39) is a 40-kDa protein that, together with the STE20-related adaptor-alpha (STRAD alpha) pseudo kinase, forms a regulatory complex capable of stimulating the activity of the LKB1 tumor suppressor protein kinase. The latter is mutated in the inherited Peutz-Jeghers cancer syndrome (PJS). CAB39 binds directly to a conserved Trp-Glu-Phe sequence at the STRAD alpha C terminus, markedly enhancing binding of STRAD alpha to LKB1 and increasing LKB1 catalytic activity. Skeletal muscle contraction results in the phosphorylation and activation of the AMP-activated protein kinase (AMPK) by an upstream kinase (AMPKK). The LKB1-STE-related adaptor (STRAD)-mouse protein 25 (MO25) complex is the major AMPKK in skeletal muscle; however, LKB1-STRAD-MO25 activity is not increased by muscle contraction. This relationship suggests that phosphorylation of AMPK by LKB1-STRAD-MO25 during skeletal muscle contraction may be regulated by allosteric mechanisms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAB39
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat