Order Entry
Export
ContactUsLinkComponent
Anti-KIF17 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-KIF17 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-3527R-A647
 :  Bioss
Anti-KIF17 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-3527R-A647
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Kinesin Family member 17
  • Antigen symbol:
    KIF17
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    57576
  • Antigen synonyms:
    KIF17B|KLP-2|KIF3X|OSM-3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

The kinesin superfamily of proteins (KIFs) consists of a class of microtubule-dependent motors that play a major role in many cellular and developmental functions, including organelle transport, mitosis, meiosis, and possibly long-range signaling in neurons. The kinesin proteins are involved in organelle transport and are primarily associated with anterograde transport of vesicles and organelles in neurons, epithelial cells, and melanosomes with bidirectional transport of mitochondria. They also mediate transport between the endoplasmic reticulum (ER) and the Golgi complex. In neurons, kinesin motors conduct vesicular transport, such as of synaptic vesicle components to axons and of neurotransmitter receptors to dendrites. KIF17 belongs to the functionally diverse subgroup of the kinesin superfamily characterized by a N-terminal motor domain (N-IV class), that includes the KIF3 motor protein. KIF17 is specifically expressed in the brain, present in abundance in the gray matter, particularly in the hippocampus and cerebral cortex, but not in the white matter such as the optic nerve.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIF17
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat