- Antibody type:Primary
- Antigen name:MLX Interacting Protein-Like
- Antigen symbol:MLXIPL
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51085
- Antigen synonyms:CHREBP|WBSCR14|WS-bHLH|bHLHd14|MIO|MONDOB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
ChREBP (Carbohydrate responsive element binding protein) is a transcription factor playing a critical role in the nutrient and hormonal regulation of genes encoding enzymes of glucose metabolism and lipogenesis pathways. It contains several domains including a nuclear localization signal (NLS) near the N-terminus, polyproline domains, a basic helix-loop-helix leucine zipper (b/HLH/Zip) and a leucine zipper like (zip-like) domain.ChREBP is ubiquitously detected in various tissues, with highest expression in liver, kidney and white and brown adipose tissue. Under basal conditions ChREBP is localized in the cytosol, translocating into the nucleus upon high glucose stimulation following its dephosphorylation of serine 196.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MLXIPL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat