Order Entry
Export
ContactUsLinkComponent
Anti-IL33 Rabbit Polyclonal Antibody (Cy7®)
Anti-IL33 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-2633R-CY7
Supplier:  Bioss
Anti-IL33 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-2633R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 33
  • Antigen symbol:
    IL33
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    77125
  • Antigen synonyms:
    NFEHEV|C9orf26|IL1F11|NF-HEV|Propeptide|DVS27
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    214-264/266
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Cytokine that binds to and signals through the IL1RL1/ST2 receptor which in turn activates NF-kappa-B and MAPK signaling pathways in target cells. Involved in the maturation of Th2 cells inducing the secretion of T-helper type 2-associated cytokines. Also involved in activation of mast cells, basophils, eosinophils and natural killer cells. Acts as a chemoattractant for Th2 cells, and may function as an "alarmin", that amplifies immune responses during tissue injury. In quiescent endothelia the uncleaved form is constitutively and abundantly expressed, and acts as a chromatin-associated nuclear factor with transcriptional repressor properties, it may sequester nuclear NF-kappaB/RELA, lowering expression of its targets. This form is rapidely lost upon angiogenic or proinflammatory activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL33
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat